missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ NucRed™ Live 647 ReadyProbes™ Reagent

Cell-permeant nuclear stain that emits bright, far-red fluorescence when bound to DNA

Brand:  Invitrogen™ R37106

220.40 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 13579180


Includes: 6 x 2.5mL dropper bottles
This item is not returnable. View return policy

Description

Description

  • Formulated as room temperature-stable solution in dropper bottle
  • Tip and drip two drops per mL to stain cells
  • Exceptionally bright far-red fluorescence well separated from blue, green, and red fluorophores
  • Rapid staining of live or fixed cells without wash steps
  • Ready-to-use liquid formulation: no need to dilute, weigh, or pipet
  • Can be used for counter-staining, DNA content measurements, cell cycle, cell replication studies, and cell counting
  • Excitation /emission of 638/686nm make it optimal for use with blue (Hoechst, DAPI, CFP), green (Alexa Fluor 488, FITC, GFP), and red (Alexa Fluor 594, Texas Red, rhodamine) fluorophores in multicolor experiments
  • May be added directly to cells in full media or buffer solutions
  • No wash, cell lysis, or RNase steps are needed
  • In most cases, 2 drops/mL and an incubation time of 15 to 30 minutes will produce bright nuclear staining; however, optimization may be needed for some cell types, conditions, and applications
  • In such cases, simply add more or fewer drops until the optimal staining intensity is obtained
  • In most cases, staining intensity increases with time if cells are not washed prior to imaging
  • Detected through far-red filters

Cell Analysis, Cell Cycle, Cell Structure, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, Flow Cytometry, Flow Cytometry Cell Cycle Assays, Flow Cytometry Staining by Cell Structure, Flow Cytometry of Cellular Processes, Fluorescence In Situ Hybridization, Immunofluorescence (IF), Immunofluorescence Counterstaining, Mounting and Fade Prevention, Immunofluorescence Staining and Detection, In Situ Hybridization (ISH), Nucleus, Nucleoli and Nuclear Envelope, Organelle Tracing

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Far-Red
638⁄686
Confocal Microscope, Fluorescence Microscope, Flow Cytometer
Fluorescence
Liquid
Fluorescent Dye
Nucleus
6 × 2.5 mL dropper bottles

Store at ≤ 25°C

Cell-Permeant
6 dropper bottle(s)
Visible
NucRed™, ReadyProbes™
Nucleic Acid Stain
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.