Learn More
Invitrogen™ Oregon Green™ 488 BAPTA-1, AM, cell permeant - Special Packaging
Cells may be loaded with the AM ester forms of these calcium indicators by adding the dissolved indicator directly to dishes containing cultured cells
Brand: Invitrogen™ O6807
394.68 EUR valid until 2024-09-28
Use promo code "21626" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”21626” to get your promotional price
Description
Labeled calcium indicators are molecules that exhibit an increase in fluorescence upon binding Ca2+. They have uses in many calcium signaling investigations, including measuring intracellular Ca2+, following Ca2+ influx and release, and multiphoton excitation imaging of Ca2+ in living tissues.
- Fluorescence signal from these cells is generally measured using fluorescence microscopy, fluorescence microplate assays, or flow cytometry
- Label (Ex/Em): Oregon Green 488 BAPTA-1 (494/523nm)
- Fluorescence intensity increase upon binding Ca2+: approximately 14 fold
- Exhibit fluorescence increase upon binding Ca2+ with little shift in wavelength
- These probes are excited by visible light, and because energy required for excitation is low, potential for cellular photodamage is reduced
- Commonly used laser-based instruments (i.e., confocal laser scanning microscopes) are able to efficiently excite these indicators, and their emissions are in regions of spectrum where cellular autofluorescence and scattering backgrounds are often less of problem
Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Cell-Based Second Messenger Assays, Drug Discovery and Development, G-Protein Coupled Receptor Biology, Ionic Homeostasis and Signaling, Target and Lead Identification and Validation
Order Info
Shipping Condition: Room temperature
Specifications
Store in freezer -5°C to -30°C. | |
Cell Viability and Proliferation | |
Stain | |
Oregon Green™ | |
Room Temperature |
Fluorescence | |
Fluorescence Microscope, Flow Cytometer, Microplate Reader | |
Fluorescent Dye-Based | |
10 x 50 μg |
For Research Use Only. Not for use in diagnostic procedures.