missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Oregon Green™ 488 BAPTA-1, Hexapotassium Salt, cell impermeant

A visible light-excitable calcium indicator

Brand:  Invitrogen™ O6806

356.44 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 11589186

  • 469.00 € / 500µg
In Stock
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy

Description

Description

  • This water-soluble, salt form is useful for intracellular loading by microinjection, infusion from patch pipet or uptake induced by Influx™ pinocytic cell-loading reagent (Cat. No. I-14402)

Calcium Detection, Cell Analysis, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
Fluorescence Microscope, Flow Cytometer, Microplate Reader
Fluorescent Dye-Based
500 μg
Fluorescence
Calcium Indicator
Oregon Green™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.