missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Paclitaxel, Oregon Green™ 488 Conjugate (Oregon Green™ 488 Taxol, Flutax-2)

Probe for labeling tubulin filaments in live cells

Brand:  Invitrogen™ P22310

576.08 EUR valid until 2025-04-18
Use promo code "24107" to get your promotional price.


Product Code. 11569226

  • 766.00 € / 100µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

The fluorescent label on this probe is attached to the 7-carbon of the paclitaxel, a strategy that permits selective binding of the probe to microtubules. Researchers have used similar fluorescent paclitaxel derivatives in a variety of systems.

Cell Analysis, Cell Structure, Cytoskeleton

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C and protect from light.
100 μg
Room Temperature
Paclitaxel
Oregon Green™ 488
Oregon Green™
Classic Dyes
Cytoskeleton, Tubulin
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2975936 Certificate TypeCertificate of Analysis Date06/07/2024 Catalog NumberP22310
Lot Number2876726 Certificate TypeCertificate of Analysis Date20/02/2024 Catalog NumberP22310
Lot Number2507888 Certificate TypeCertificate of Analysis Date01/08/2022 Catalog NumberP22310
Lot Number2335661 Certificate TypeCertificate of Analysis Date13/09/2021 Catalog NumberP22310
Lot Number2157077 Certificate TypeCertificate of Analysis Date28/11/2019 Catalog NumberP22310
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Paclitaxel, Oregon Green™ 488 Conjugate (Oregon Green™ 488 Taxol, Flutax-2) > 100ug

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.