Learn More
Invitrogen™ PED6 (N-((6-(2,4-Dinitrophenyl)amino)hexanoyl)-2-(4,4-Difluoro-5,7-Dimethyl-4-Bora-3a,4a-Diaza-s-Indacene-3-Pentanoyl)-1-Hexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt)
PED6-labeled phospholipid
Brand: Invitrogen™ D23739
590.90 EUR valid until 2025-02-21
Use promo code "24115" to get your promotional price.
Description
Molecular Probes phospholipase substrates are dye-labeled phospholipids used to monitor phospholipase A (PLA) activity in purified enzyme preparations, cell lysates, and living cells.
- Label (Ex/Em): PED6
- Specificity: PLA2 (this substrate is cleaved by PLA1, but this reaction does not separate the fluorophore from the quencher)
- Cleavage product: BODIPY™ FL C5
- Lyophilized product can be dissolved in DMSO for use
- Fluorescence is measured either in microplate assays or using flow cytometry
Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Cellular Imaging, Immunofluorescence (IF), Immunofluorescence Staining and Detection
Order Info
Shipping Condition: Room temperature
Specifications
Phospholipids | |
Other Labels or Dyes | |
Store in freezer (-5 to -30°C) and protect from light. | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.