missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Phospholipase C Protein, Phosphatidylinositol-Specific (from Bacillus cereus)

Suitable for enzymology and cell biology applications

Brand:  Invitrogen™ P6466

178.50 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10615763

  • 255.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

This phosphatidylinositol-specific phospholipase C is a highly purified enzyme preparation from Bacillus cereus which cleaves phospholipid phosphatidylinositol (PI) into two molecules. Validated for assay standard. Alphabetical protein filter: P

Cell Analysis, Cell Structure, Enzyme & Protein Activity Assays, Lipase & Phopholipase Activity, Membranes (General) & Lipids, Plasma Membrane, Proteins, Expression, Isolation & Analysis

Order Info

Shipping Condition: Wet Ice

TRUSTED_SUSTAINABILITY
Specifications

Specifications

100 U⁄ml
50 μL
B. cereus
Wet Ice
Other Proteins
Assay Standard
Bacillus cereus
Store in freezer (-5 to -30°C).
Solution
Signal Transduction
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2892430 Certificate TypeCertificate of Analysis Date15/03/2024 Catalog NumberP6466
Lot Number2541665 Certificate TypeCertificate of Analysis Date09/03/2023 Catalog NumberP6466
Lot Number2331899 Certificate TypeCertificate of Analysis Date26/03/2021 Catalog NumberP6466
Lot Number2273750 Certificate TypeCertificate of Analysis Date20/10/2020 Catalog NumberP6466
Lot Number2030244 Certificate TypeCertificate of Analysis Date29/09/2020 Catalog NumberP6466
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Phospholipase C Protein, Phosphatidylinositol-Specific (from Bacillus cereus) > 50uL

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.