missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Pro-Q™ Emerald 300 Glycoprotein Gel and Blot Stain Kit

Product Code. 10195002
Click to view available options
Quantity:
1 kit
Unit Size:
9g
This item is not returnable. View return policy

Product Code. 10195002

Brand: Invitrogen™ P21857

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

50-fold more sensitive than standard fuchsin staining

Pro-Q Emerald 300 and Pro-Q Emerald 488 Glycoprotein Stain Kits provide the most advanced technology available for detection of glycoproteins in gels and on blots. Gel staining is rapid and very sensitive. In less than three hours, it is possible to detect as little as 300 pg of glycoprotein per band, depending on the degree of glycosylation.

Protein Gel Electrophoresis, Protein Gel Staining and Imaging, Proteins, Expression, Isolation and Analysis

TRUSTED_SUSTAINABILITY

Specifications

Detection Location In-Blot Detection, In-Gel Detection
Detection Method Fluorescence
Label or Dye Pro-Q Emerald 300
Product Type Glycoprotein Staining Kit
Product Line PRO-Q
Quantity 1 kit
Shipping Condition Room Temperature
Target Molecule Proteins (Glycoproteins)

For Research Use Only. Not for use in diagnostic procedures.

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.