missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ 5-(and-6)-Carboxy SNARF™-1

The cell-impermeant pH indicator has pKa of approximately 7.5, thus is useful for measuring pH changes between pH 7 and pH 8

Brand:  Invitrogen™ C1270

459.04 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.


Product Code. 10779394

  • 604.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • Exhibits significant pH-dependent emission shift from yellow-orange to deep red fluorescence under acidic and basic conditions, respectively
  • This pH dependence allows ratio of fluorescence intensities from dye at two emission wavelengths, typically 580nm and 640nm, to be used for quantitative determinations of pH

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature and protect from light.
Fluorescence Microscope, Flow Cytometer, Microplate Reader
pH Indicator
1 mg
Fluorescence
pH Indicator
SNARF™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

For Research Use Only. Not for use in diagnostic procedures.