missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ SNARF™-5F 5-(and-6)-Carboxylic Acid, Acetoxymethyl Ester, Acetate - Special Packaging

Cell-permeant ratiometric pH indicator

Brand:  Invitrogen™ S23923

Product Code. 11590666

  • 752.00 € / 1000µg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • pKa of approximately 7.2
  • Exhibits a significant pH-dependent emission shift from yellow-orange to deep red fluorescence under acidic and basic conditions, respectively
  • This pH dependence allows the ratio of the fluorescence intensities from the dye at two emission wavelengths, typically 580nm and 640nm, to be used for quantitative determinations of pH

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, pH Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store in freezer -5°C to -30°C.
Fluorescence Microscope, Flow Cytometer, Microplate Reader
Fluorescent Dye-Based
SNARF™
Room Temperature
Fluorescence
pH Indicator
pH Indicator
20 x 50 μg
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.