missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Sodium Green™, Tetra (Tetramethylammonium) Salt, cell impermeant

Visible light-excitable Na+ indicator

Brand:  Invitrogen™ S6900

345.04 EUR valid until 2024-12-31
Use promo code "21651" to get your promotional price.


Product Code. 10645773

  • 454.00 € / 1mg

Please to purchase this item. Need a web account? Register with us today!

Explore more special offers
This item is not returnable. View return policy

Description

Description

  • For use with fluorescence microscope and flow cytometer
  • Fluorescent dye-based with sodium indicator

Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Ionic Homeostasis and Signaling, Sodium and Potassium Detection

Order Info

Shipping Condition: Room temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Store at room temperature and protect from light.
Fluorescence Microscope, Flow Cytometer
Sodium Indicator
1 mg
Fluorescence
Sodium Indicator
Sodium Green™
Room Temperature
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.