missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Tali™ Cell Cycle Kit
Convenient and accurate determination of the percentage of cells in each phase of the cell cycle
Brand: Invitrogen™ A10798
91.57 EUR valid until 2024-12-31
Use promo code "21956" to get your promotional price.
Includes: 1 bottle
Description
The TaliCell Cycle Kit includes a ready-to-use reagent that can be used with the TaliImage-Based Cytometer. This single-addition reagent is composed of propidium iodide (PI), RNase A, and Triton X-100.
The TaliCell Cycle Kit offers a simple workflow:
- Pre-mixed reagent can be stored at room temperature and used when your cells are ready
- Simply fix your cells, add the reagent, incubate, and visualize
- Cell cycle data can be analyzed on the Taliinstrument or exported as a .csv or .fcs file for analysis with various modeling software
Cell Analysis, Cell Tracing and Tracking, Cell Viability, Proliferation and Function, Cellular Imaging, DNA Replication and Repair, General Cell Tracing, Imaging Cytometry
Specifications
Cell Cycle Kit | |
Fluorescence | |
1 bottle, store at room temperature. | |
Mammalian Cells | |
Bottle | |
50 Assays |
Mammalian Cell | |
Tali™ Image-Based Cytometer, Tali™ Image-Based Cytomoter | |
Red | |
Propidium Iodide | |
Tali™ |
For Research Use Only. Not for use in diagnostic procedures.