missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Texas Red™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (Texas Red™ DHPE)

The phospholipid, Texas Red™ 1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt (Texas Red™ DHPE) is labeled on the head group with the bright, red-fluorescent Texas Red™ dye.

Brand:  Invitrogen™ T1395MP

261.10 EUR valid until 2025-06-30
Use promo code "24074" to get your promotional price.


Product Code. 10338022

  • 373.00 € / Each

Please to purchase this item. Need a web account? Register with us today!

Edge
Explore more special offers
This item is not returnable. View return policy

Description

Description

The phospholipid, Texas Red™ 1,2-dihexadecanoyl-sn-glycero-3-phosphoethanolamine, triethylammonium salt (Texas Red™ DHPE) is labeled on the head group with the bright, red-fluorescent Texas Red™ dye (excitation/emission maxima ∼595/615 nm).

To make a concentrated stock solution from 1–2 mg/mL, dissolve in 100% ethanol. Sonication may be necessary to aid dispersion. Store frozen, desiccated and protected from light. The gradual accumulation of moisture from the atmosphere may promote aggregation of the reagent over time.

Order Info

Shipping Condition: Room Temperature

TRUSTED_SUSTAINABILITY
Specifications

Specifications

Phospholipids
1381.85
Texas Red
1 mg
Solid
583/601 nm
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number2946763 Certificate TypeCertificate of Analysis Date22/08/2024 Catalog NumberT1395MP
Lot Number2770861 Certificate TypeCertificate of Analysis Date23/10/2023 Catalog NumberT1395MP
Lot Number2497579 Certificate TypeCertificate of Analysis Date07/09/2022 Catalog NumberT1395MP
Lot Number2369105 Certificate TypeCertificate of Analysis Date24/07/2021 Catalog NumberT1395MP
Lot Number2274776 Certificate TypeCertificate of Analysis Date03/12/2020 Catalog NumberT1395MP
Special Offers

Special Offers

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Invitrogen™ Texas Red™ 1,2-Dihexadecanoyl-sn-Glycero-3-Phosphoethanolamine, Triethylammonium Salt (Texas Red™ DHPE) > 1 mg

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only. Not for use in diagnostic procedures.