Learn More
Invitrogen™ Thiol and Sulfide Quantitation Kit
Ultrasensitive colorimetric assay for quantifying both protein and nonprotein thiols
Brand:  Invitrogen™ T6060
414.96 EUR valid until 2024-12-20
Use promo code "21651" to get your promotional price.
Product Code. 10388252
  • 546.00 € / Each
Estimated Shipment: 13-12-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
In this assay, thiols or inorganic sulfides reduce a disulfide-inhibited derivative of papain, stoichiometrically releasing the active enzyme. The activity of the enzyme is then measured using the chromogenic papain substrate N-benzoyl-L-arginine, p-nitroanilide (L-BAPNA). The enzymatic amplification step in this kit enables researchers to detect as little as 0.2μM thiol - a sensitivity that is about 100-fold greater than that achieved using Ellman's reagent.
Anion Detection, Biochemical Toxicology Assays, Cell Analysis, Cell Metabolism, Cell Viability, Proliferation and Function, Drug Discovery and Development, Ionic Homeostasis and Signaling, Target-Based ADME/Tox Assays
Order Info
Shipping Condition: Room temperature
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Thiol and Sulfide Quantitation Kit
1 kit