Learn More
Invitrogen™ ViaGram™ Red+ Bacterial Gram Stain and Viability Kit
Description
The ViaGram Red+ Bacterial Gram Stain and Viability Kit differentially stains many gram-positive and gram-negative bacterial species and, at the same time, discriminates live from dead cells on the basis of plasma membrane integrity.
Analysis of Bacteria, Bacterial Viability & Cytotoxicity Analysis, Cell Analysis, Cell Viability & Cytotoxicity, Cell Viability, Proliferation & Function, Cellular Imaging, Gram Staining, Immunofluorescence (IF), Immunofluorescence Staining & Detection, Microbiological Analysis
Informations sur la commande
Shipping Condition: Room Temperature
Spécification
Spécification
| Content And Storage | Store in freezer (-5 to -30°C) and protect from light. |
| Detection Method | Fluorescence |
| For Use With (Equipment) | Fluorescence Microscope |
| Quantity | 1 Kit |
| Shipping Condition | Room Temperature |
| Conjugate | Other Labels or Dyes |
| Format | Liquid |
For Research Use Only. Not for use in diagnostic procedures.
Veuillez fournir vos retours sur le contenu du produit en remplissant le formulaire ci-dessous.