Learn More
Invitrogen™ Vybrant™ Cell Metabolic Assay Kit, with C12-resazurin
C12-resazurin reduced to C12-resorufin
Marca: Invitrogen™ V23110
Descripción
In the Vybrant™ Cell Metabolic Assay Kit, nonfluorescent C12-resazurin is reduced to fluorescent C12-resorufin by viable cells. The resulting signal is proportional to the number of cells present. C12-resazurin has better cellular retention than alamarBlue™ dye and can be multiplexed with viability indicators and other biomarkers.
- Platform: Flow Cytometer, Microplate Reader
- Detection Method: Fluorescent
- Format: Tube(s), 96-well plate
Cell Analysis, Cell Metabolism, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function, Cellular Imaging, High-Content Screening (HCS)
Información de órdenes
Shipping Condition: Room temperature
Especificaciones
Store in freezer -5°C to -30°C and protect from light. | |
Cell Metabolic Assay | |
Dye | |
Tube(s), 96-well plate | |
1 kit |
Fluorescence | |
Flow Cytometer, Microplate Reader | |
Resorufin | |
Vybrant™ | |
Room Temperature |
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
For Research Use Only. Not for use in diagnostic procedures.