Learn More
Invitrogen™ CyQUANT™ Cytotoxicity Assay Kit (G6PD Release Assay)
Detect as few as 500 cells.
Brand: Invitrogen™ V23111
533.36 EUR valid until 2025-03-21
Use promo code "24111" to get your promotional price.
Alert:
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”24111” to get your promotional price
Description
In the Vybrant™ Cytotoxicity Assay Kit, damaged and dying cells release glucose 6-phosphate dehydrogenase into surrounding medium. The glucose 6-phosphate dehydrogenase is detected by an enzymatic process that leads to the reduction of resazurin into red-fluorescent resorufin. This assay can detect as few as 500 cells and is more sensitive than LDH release assays.
Cell Analysis, Cell Viability and Cytotoxicity, Cell Viability, Proliferation and Function
Order Info
Shipping Condition: Room temperature

Specifications
Store in freezer -5°C to -30°C and protect from light. | |
Cytotoxicity Assay Kit (G6PD Release Assay) | |
Stain | |
96-well plate | |
1 kit |
Fluorescence | |
Microplate Reader | |
Other Label(s) or Dye(s) | |
CyQUANT™ | |
Room Temperature |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
For Research Use Only. Not for use in diagnostic procedures.