missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieve Deperox/Dehydrate, Honeywell Fluka™
Beads, for the deperoxidation and drying of solvents
228.03 € - 738.34 €
Specifications
Packaging | Poly bottle |
---|---|
Chemical Name or Material | Molecular Sieve Deperox/Dehydrate Fluka for the deperoxidation and drying of solvents |
CAS | 10124-43-3, 1318-02-1 |
UN Number | UN3077 |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Poly bottle | |
Molecular Sieve Deperox/Dehydrate Fluka for the deperoxidation and drying of solvents | |
10124-43-3, 1318-02-1 | |
UN3077 |
Safety and Handling
P201-P261-P280-P284-P304 + P340-P308 + P313
missing translation for 'dotInformation' : 9/PGIII