missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 13X, Honeywell™ Fluka™
Shop All Honeywell Fluka Products
Click to view available options
Quantity:
1 kg
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Specifications
Specifications
Packaging | Poly Bottle |
Quantity | 1 kg |
CAS | 63231-69-6 |
Linear Formula | Na86[AlO2)86(SiO2)106] · xH2O |
MDL Number | MFCD00131613 |
UN Number | NONH for all modes of transport |
IUPAC Name | Molecular sieves 13X |
Chemical Name or Material | Molecular Sieves, 13X |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction