missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads, Thermo Scientific Chemicals

Product Code. 11387455
Click to view available options
Quantity:
250 g
1000 g
5000 g

Molecular sieves, 4A, 3-5 mm (0.12-0.20in) beads are useful in general gas drying applications.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
MDL Number MFCD00131613
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Sensitivity Hygroscopic
Quantity 1000 g
Solubility Information Insoluble in cold water,hot water,methanol,diethyl ether,n-octanol and acetone.
Chemical Name or Material Molecular sieves, 4A, 3 to 5mm (0.12 to 0.20 in) beads

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'