missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 4A, powder <50 micron, Thermo Scientific Chemicals

Product Code. 10698124
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
10 kg
Packaging:
Plastic Bottle
Plastic drum
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 70955-01-0
Molecular Formula C20H25FN2O8
Molecular Weight (g/mol) 440.42
InChI Key FJUZEZRZKVMAMD-UHFFFAOYNA-N
PubChem CID 73906282
IUPAC Name (2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid
SMILES COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC

Specifications

Color White to Beige
pH 9 to 12 (10% water suspension)
Packaging Plastic Bottle
Residual Water 3% max.
Water Adsorption 18% max.
Quantity 2.5 kg
Physical Form Powder
MDL Number MFCD00131613
Solubility Information Solubility in water: insoluble.
Chemical Name or Material Molecular sieves 4A, Powder, <50 micron

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'