missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molecular Sieves, 4A, Powder, Activated, −325 mesh, Honeywell Fluka™
Powder, activated, −325 mesh particle size
51.46 € - 339.96 €
Chemical Identifiers
CAS | 70955-01-0 |
---|---|
Molecular Formula | C20H25FN2O8 |
Molecular Weight (g/mol) | 440.42 |
MDL Number | MFCD00131613 |
InChI Key | FJUZEZRZKVMAMD-UHFFFAOYNA-N |
PubChem CID | 73906282 |
SMILES | COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15646710
|
Honeywell-Fluka
688363-25G |
25 g | N/A | ||||||
15656710
|
Honeywell-Fluka
688363-500G |
500 g | N/A | ||||||
15626710
|
Honeywell-Fluka
688363-1KG |
1 kg | N/A | ||||||
15636710
|
Honeywell-Fluka
688363-2.5KG |
2.5 kg | N/A | ||||||
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
70955-01-0 | |
440.42 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC |
C20H25FN2O8 | |
MFCD00131613 | |
73906282 |
Specifications
Tan | |
Poly bottle | |
Powder, activated | |
C20H25FN2O8 | |
MFCD00131613 | |
FJUZEZRZKVMAMD-UHFFFAOYNA-N | |
(2R,3S)-2-[[(2R)-2-acetamido-3-(3-fluorophenyl)propanoyl]amino]-5-methoxy-4-methoxycarbonyl-3-methyl-5-oxopentanoic acid | |
73906282 |
2g/cm3 | |
70955-01-0 | |
400°C | |
Na12[(AlO2)12(SiO2)12] · xH2O | |
NONH for all modes of transport | |
COC(=O)C(C(C)C(NC(=O)C(CC1=CC(F)=CC=C1)NC(C)=O)C(O)=O)C(=O)OC | |
440.42 | |
Molecular sieves, 4A |
Safety and Handling
P261-P305 + P351 + P338