missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves, 5A, 3-5mm (0.12-0.20in) beads, Thermo Scientific Chemicals

Product Code. 11327575
Click to view available options
Quantity:
250 g
1000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Molecular sieves, 13X, 3-5mm (0.12-0.20in) beads are used for liquefied petroleum gas, natural gas drying, desulfurization, clean. It is used as an adsorbent for gases and liquids. It is used in chemistry laboratory for separating compounds and drying reaction starting materials.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69912-79-4
Molecular Formula AlNaO6Si2
Molecular Weight (g/mol) 202.14
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
PubChem CID 19758701
IUPAC Name aluminum;sodium;dioxido(oxo)silane
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O

Specifications

Sensitivity Hygroscopic
Quantity 250 g
Solubility Information Insoluble in water.
Chemical Name or Material Molecular sieves, 5A, 3 to 5mm (0.12 to 0.20 in) beads

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'