missing translation for 'onlineSavingsMsg'
Learn More

Molecular sieves 5A, powder <50 micron, Thermo Scientific Chemicals

Product Code. 10296980
Click to view available options
Quantity:
500 g
Packaging:
Glass bottle

 

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Drying agent
TRUSTED_SUSTAINABILITY

Specifications

Color White
pH 10 to 10.8 (10% aq. suspension)
Packaging Glass bottle
Residual Water < 1.5% (550°C, 2 hrs.)
Water Adsorption > 23% (50%rH, 20°C, 24h)
Quantity 500 g
CAS 69912-79-4
Physical Form Fine Powder
Molecular Formula AlNaO6Si2
MDL Number MFCD00147627
InChI Key URGAHOPLAPQHLN-UHFFFAOYSA-N
SMILES [Na+].[Al+3].[O-][Si]([O-])=O.[O-][Si]([O-])=O
IUPAC Name aluminum;sodium;dioxido(oxo)silane
Molecular Weight (g/mol) 202.14
PubChem CID 19758701
Chemical Name or Material Molecular sieves 5A
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'