missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum boride, 99% (metals basis), Thermo Scientific Chemicals

81.20 € - 268.34 €

Chemical Identifiers

CAS 12006-98-3
Molecular Formula BMo
Molecular Weight (g/mol) 106.76
MDL Number MFCD00014219
InChI Key LGLOITKZTDVGOE-UHFFFAOYSA-N
Synonym molybdenum boride, molybdenum boride mob, molybdenumylidyneborane, molybdenum monoboride, molybdenum boride mono , mob, molybdenum boride, powder,-325 mesh trace metals basis
PubChem CID 82790
IUPAC Name boranylidynemolybdenum
SMILES B#[Mo]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11367407
View Documents
Thermo Scientific Alfa Aesar
012563.14
25 g
81.20 €
25g
Please to purchase this item. Need a web account? Register with us today!
11347407
View Documents
Thermo Scientific Alfa Aesar
012563.22
100 g
268.34 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Molybdenum borides have been used as hard coating materials for high temperature structural applications, and also in the form of MoB/CoCr cermet coatings. It is also used as a brazing material especially in electric components. Molybdenum boride has potential industrial applications such as abrasive, corrosion resistant.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

12006-98-3
106.76
LGLOITKZTDVGOE-UHFFFAOYSA-N
82790
B#[Mo]
BMo
MFCD00014219
molybdenum boride, molybdenum boride mob, molybdenumylidyneborane, molybdenum monoboride, molybdenum boride mono , mob, molybdenum boride, powder,-325 mesh trace metals basis
boranylidynemolybdenum

Specifications

12006-98-3
BMo
molybdenum boride, molybdenum boride mob, molybdenumylidyneborane, molybdenum monoboride, molybdenum boride mono , mob, molybdenum boride, powder,-325 mesh trace metals basis
LGLOITKZTDVGOE-UHFFFAOYSA-N
boranylidynemolybdenum
82790
99%
Molybdenum boride
Powder
MFCD00014219
it is soluble in water,paraffin oil,ether,benzene and other organic solvents.
B#[Mo]
106.76
106.75
(metals basis)
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 234-492-5

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.