missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum carbide, 99.5% (metals basis), Thermo Scientific Chemicals
125.00 € - 552.00 €
Chemical Identifiers
CAS | 12069-89-5 |
---|---|
Molecular Formula | CMo2 |
Molecular Weight (g/mol) | 203.91 |
MDL Number | MFCD00014218 |
InChI Key | QIJNJJZPYXGIQM-UHFFFAOYSA-N |
Synonym | dimolybdenum carbide, methane dimolybdenum, molybdenum carbide mo2c, molybdenum carbide trace metals basis 100g |
IUPAC Name | (molybdeniomethylidene)molybdenum |
SMILES | [Mo]=C=[Mo] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11316588
|
Thermo Scientific Alfa Aesar
012192.22 |
100 g |
125.00 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11306588
|
Thermo Scientific Alfa Aesar
012192.36 |
500 g |
552.00 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molybdenum carbide is used in the production of solid molybdenum carbide grades of hard metal in the manufacture of mixed crystal. It used as coating material.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
12069-89-5 | |
203.91 | |
QIJNJJZPYXGIQM-UHFFFAOYSA-N | |
(molybdeniomethylidene)molybdenum |
CMo2 | |
MFCD00014218 | |
dimolybdenum carbide, methane dimolybdenum, molybdenum carbide mo2c, molybdenum carbide trace metals basis 100g | |
[Mo]=C=[Mo] |
Specifications
White | |
2687°C | |
8.9g/cm3 at 20°C | |
CMo2 | |
dimolybdenum carbide, methane dimolybdenum, molybdenum carbide mo2c, molybdenum carbide trace metals basis 100g | |
QIJNJJZPYXGIQM-UHFFFAOYSA-N | |
(molybdeniomethylidene)molybdenum | |
203.91 | |
Powder |
12069-89-5 | |
White powder | |
99.5% (metals basis) | |
MFCD00014218 | |
Insoluble in water. | |
[Mo]=C=[Mo] | |
203.91 | |
Odorless | |
Molybdenum carbide |
Safety and Handling
missing translation for 'einecsNumber' : 235-115-7
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only