missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum gauze, 20 mesh woven from 0.305mm (0.012 in.) dia. wire, Thermo Scientific Chemicals
Filtration, electrodes
74.77 € - 732.00 €
Chemical Identifiers
CAS | 7439-98-7 |
---|---|
Molecular Formula | Mo |
Molecular Weight (g/mol) | 95.95 |
MDL Number | MFCD00003465 |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
Synonym | molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 |
PubChem CID | 23932 |
ChEBI | CHEBI:28685 |
IUPAC Name | molybdenum |
SMILES | [Mo] |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11355187
|
Thermo Scientific Alfa Aesar
040908.FL |
75 x 75 mm |
74.77 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11375187
|
Thermo Scientific Alfa Aesar
040908.GT |
150 x 150 mm |
218.09 €
Each |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11365187
|
Thermo Scientific Alfa Aesar
040908.HB |
300 x 300 mm | N/A | ||||||
Description
Filtration, electrodes
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
CHEBI:28685 | |
[Mo] |
Specifications
7439-98-7 | |
MFCD00003465 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
20 Mesh | |
Molybdenum gauze, Woven from 0.305mm (0.012 in.) dia. wire |
Mo | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
Wire Cloth |
Safety and Handling
missing translation for 'einecsNumber' : 231-107-2
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only