Learn More
Molybdenum(III) chloride, 99.5% (metals basis), Thermo Scientific Chemicals
91.60 € - 360.00 €
Chemical Identifiers
CAS | 13478-18-7 |
---|---|
Molecular Formula | Cl3Mo |
Molecular Weight (g/mol) | 202.3 |
MDL Number | MFCD00014214 |
InChI Key | ZSSVQAGPXAAOPV-UHFFFAOYSA-K |
Synonym | molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride |
PubChem CID | 83515 |
ChEBI | CHEBI:30626 |
IUPAC Name | trichloromolybdenum |
SMILES | Cl[Mo](Cl)Cl |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11321569
|
Thermo Scientific Alfa Aesar
014034.04 |
2 g |
91.60 €
2g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
11331569
|
Thermo Scientific Alfa Aesar
014034.09 |
10 g |
360.00 €
10g |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Molybdenum(III) chloride reacts with tetrahydrothiophene to form complexes, which catalyze the polymerization and cyclotrimerization of alkynes.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
13478-18-7 | |
202.3 | |
ZSSVQAGPXAAOPV-UHFFFAOYSA-K | |
83515 | |
trichloromolybdenum |
Cl3Mo | |
MFCD00014214 | |
molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride | |
CHEBI:30626 | |
Cl[Mo](Cl)Cl |
Specifications
(decomposition) | |
Red | |
Cl3Mo | |
UN3260 | |
Soluble in nitric acid and sulfuric acid. Insoluble in water and dilute hydrochloric acid. Slightly soluble in ethanol and diethyl ether. | |
Cl[Mo](Cl)Cl | |
202.3 | |
CHEBI:30626 | |
99.5% | |
Air and moisture sensitive | |
Molybdenum(III) chloride |
13478-18-7 | |
Powder | |
MFCD00014214 | |
molybdenum trichloride, molybdenum iii chloride, molybdenum chloride mocl3, mocl3, trichloridomolybdenum, acmc-1bwlx, molybdenum 3+ chloride | |
ZSSVQAGPXAAOPV-UHFFFAOYSA-K | |
trichloromolybdenum | |
83515 | |
202.32 | |
Acidic | |
(metals basis) |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c
H314-H335
missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.
missing translation for 'einecsNumber' : 236-775-9
missing translation for 'rtecsNumber' : QA4681400
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures; Store under Argon
RUO – Research Use Only