missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(IV) oxide, 99% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 18868-43-4
Molecular Formula MoO2
Molecular Weight (g/mol) 127.948
MDL Number MFCD00014223
InChI Key QXYJCZRRLLQGCR-UHFFFAOYSA-N
Synonym molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
PubChem CID 29320
IUPAC Name dioxomolybdenum
SMILES O=[Mo]=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11408640
View Documents Promotion Details
Thermo Scientific Alfa Aesar
048117.14
25 g N/A
N/A
11498630
View Documents Promotion Details
Thermo Scientific Alfa Aesar
048117.22
100 g N/A
N/A
Description

Description

Molybdenum(IV) oxide is used to catalyze the dehydrogenation of alcohols. It is also used as an intermediates and processing aids. It finds application in the manufacture of molybdenum nano-wires.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

18868-43-4
127.948
QXYJCZRRLLQGCR-UHFFFAOYSA-N
29320
O=[Mo]=O
MoO2
MFCD00014223
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
dioxomolybdenum

Specifications

18868-43-4
Powder
MoO2
molybdenum dioxide, molybdenum iv oxide, molybdenum oxide, molybdenum iv oxide trace metals basis 25g, mm4
O=[Mo]=O
127.948
127.94
Odorless
(metals basis)
Brown to Black
99% (metals basis)
MFCD00014223
QXYJCZRRLLQGCR-UHFFFAOYSA-N
dioxomolybdenum
29320
99%
6.47 g/mL
Molybdenum (IV) oxide
Videos
Safety and Handling

Safety and Handling

In case of contact with eyes, rinse immediately with plenty of water and seek medical advice. Wear suitable protective clothing and gloves. In case of insufficient ventilation, wear suitable respiratory equipment.

H301-H332-H319-H335: Toxic if swallowed. Harmful if inhaled. Causes serious eye irritation. May cause respiratory irritation.

missing translation for 'einecsNumber' : 242-637-9

missing translation for 'rtecsNumber' : QA4688000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only