Learn More
Molybdenum(IV) selenide, 99.9% (metals basis), Thermo Scientific Chemicals
Description
Molybdenum(IV) selenide is used in transistors and photodetectors. It is also used as a solid lubricant. Further, it is used in piezoelectric and solar cells.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Specifications
Melting Point | >1200°C |
Color | Gray |
Physical Form | Powder |
Quantity | 25 g |
Assay Percent Range | 99.9% (metals basis) |
Solubility Information | Insoluble in water. |
Formula Weight | 253.87 |
Odor | Odorless |
Appearance | Dark gray powder |
Chemical Name or Material | Molybdenum(IV) selenide |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.