Learn More
Molybdenum(IV) sulfide, 98%, Thermo Scientific Chemicals
Hydrogenation and Isomerization Catalyst, Dry Lubricant
Chemical Identifiers
CAS | 1317-33-5 |
---|---|
Molecular Formula | MoS2 |
Molecular Weight (g/mol) | 160.07 |
MDL Number | MFCD00003470 |
InChI Key | CWQXQMHSOZUFJS-UHFFFAOYSA-N |
Synonym | molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z |
PubChem CID | 14823 |
ChEBI | CHEBI:30704 |
SMILES | S=[Mo]=S |
Description
It is widely used as a solid lubricant .It finds applications mainly as Intermediates, Process regulators, Processing aids. Employed as a co catalyst for desulfurization in petrochemistry. As a catalyst in the electrolysis of water.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1317-33-5 | |
160.07 | |
CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
14823 | |
S=[Mo]=S |
MoS2 | |
MFCD00003470 | |
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
CHEBI:30704 |
Specifications
1185°C | |
Black | |
98% | |
MFCD00003470 | |
Insoluble in water and dilute acids | |
S=[Mo]=S | |
160.07 | |
CHEBI:30704 | |
Odorless | |
Molybdenum(IV) sulfide |
1317-33-5 | |
Powder | |
MoS2 | |
molybdenum disulfide, molybdenite, molybdenum iv sulfide, molybdenum disulphide, molysulfide, molykote, motimol, nichimoly c, sumipowder pa, molykote z | |
CWQXQMHSOZUFJS-UHFFFAOYSA-N | |
disulfanylidenemolybdenum | |
14823 | |
160.08 | |
Black powder |
Safety and Handling
P261-P271-P304+P340-P312
H332
missing translation for 'einecsNumber' : 215-263-9
missing translation for 'rtecsNumber' : QA4697000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
- Molybdenum(IV) sulfide
Signal Word
- Warning
Hazard Category
- Acute toxicity Category 4
Hazard Statement
- H332-Harmful if inhaled.
Precautionary Statement
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
Supplemental information
- MIXTURE LIST-Contains : Molybdenum (IV) sulfide
RUO – Research Use Only