missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum plate, 12.5mm (0.492 in.) thick, 99.95% (metals basis), Thermo Scientific™

Product Code. 11315739
Click to view available options
Quantity:
50 x 50 mm
50 x 200 mm
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]

Specifications

Quantity 50 x 50 mm
Percent Purity 99.95%
Weight ≈319g/50x50mm
Form Plate
Assay (metals basis)
Chemical Name or Material Molybdenum plate, 12.5mm (0.492 in.) thick
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'