missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum powder, -100 mesh, 99.95% (metals basis), Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7439-98-7 |
---|---|
Molecular Formula | Mo |
Molecular Weight (g/mol) | 95.95 |
MDL Number | MFCD00003465 |
InChI Key | ZOKXTWBITQBERF-UHFFFAOYSA-N |
Synonym | molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 |
PubChem CID | 23932 |
ChEBI | CHEBI:28685 |
IUPAC Name | molybdenum |
SMILES | [Mo] |
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
7439-98-7 | |
95.95 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
23932 | |
molybdenum |
Mo | |
MFCD00003465 | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
CHEBI:28685 | |
[Mo] |
Specifications
7439-98-7 | |
99.95% (metals basis) | |
MFCD00003465 | |
ZOKXTWBITQBERF-UHFFFAOYSA-N | |
molybdenum | |
23932 | |
95394 | |
Powder |
10.22 | |
Mo | |
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2 | |
[Mo] | |
95.95 | |
CHEBI:28685 | |
-100 Mesh | |
Molybdenum |
Safety and Handling
P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c
H228-H360-H371
missing translation for 'dotInformation' : Hazard Class: 4.1; Packaging Group: II
missing translation for 'einecsNumber' : 231-107-2
missing translation for 'rtecsNumber' : QA4680000
missing translation for 'tsca' : Yes
Recommended Storage : Ambient temperatures
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Molybdenum powder
Signal Word
- Warning
Hazard Category
- Flammable solid Category 2
Hazard Statement
- H228-Flammable solid.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P240-Ground/bond container and receiving equipment.
- P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
- P308+P313-IF exposed or concerned: Get medical advice/attention.
- P370+P378-In case of fire: Use to extinguish.
RUO – Research Use Only