missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum powder, APS 3-7 micron, 99.95% (metals basis), Thermo Scientific Chemicals

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]
View More Specs

Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]

Specifications

Gray
gray powder
99.95% (Metals basis)
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
Odorless
Molybdenum
7439-98-7
10.22g/cm3 at 20°C
Mo
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685
Powder
Videos
Safety and Handling

Safety and Handling

P201-P202-P210-P240-P241-P260-P264b-P270-P281-P308+P313-P370+P378q-P501c

H228-H360-H371

missing translation for 'dotInformation' : DOT Class: 4.1; Packaging Group: II

missing translation for 'einecsNumber' : 231-107-2

missing translation for 'rtecsNumber' : QA4680000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

Product Identifier
  • Molybdenum powder

Signal Word
  • Warning

Hazard Category
  • Flammable solid Category 2

Hazard Statement
  • H228-Flammable solid.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P370+P378-In case of fire: Use fire-fighting equipment on basis class D to extinguish.

Supplemental information
  • MIXTURE LIST-Contains : Molybdenum

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only