Learn More
Molybdenum(V) isopropoxide, 99.6% (m.b.), 5% w/v in isopropanol, packaged under Argon in resealable ChemSeal™ bottles, Thermo Scientific Chemicals
Description
Used in the preparations of nanomaterials and thin films.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
- Packaged under argon

Specifications
Specifications
Color | Yellow to Brown |
Flash Point | 12°C (54°F) |
Odor | Alcohol-like |
Packaging | ChemSeal™ bottles |
Linear Formula | Mo[OCH(CH3)2]5 |
Quantity | 25 mL |
UN Number | UN1219 |
Sensitivity | Moisture sensitive |
Solubility Information | Soluble in hydrocarbons,reacts with alcohols,ketones and esters |
Formula Weight | 391.04 |
Show More |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
Y11G018 | Certificate of Analysis | 09/05/2023 | 041836.AC |
W24B023 | Certificate of Analysis | 08/12/2021 | 041836.AC,041836.AE |
C18N20 | Certificate of Analysis | 07/12/2021 | 041836.AC,041836.AE |
A29S058 | Certificate of Analysis | 07/12/2021 | 041836.AC,041836.AE |
H26Z033 | Certificate of Analysis | 07/12/2021 | 041836.AC,041836.AE |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.