missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(VI) oxide, 99.5+%, ACS reagent, Thermo Scientific Chemicals
50.75 € - 167.84 €
Chemical Identifiers
CAS | 1313-27-5 |
---|---|
Molecular Formula | MoO3 |
Molecular Weight (g/mol) | 143.95 |
MDL Number | MFCD00003469 |
InChI Key | JKQOBWVOAYFWKG-UHFFFAOYSA-N |
Synonym | molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide |
PubChem CID | 14802 |
ChEBI | CHEBI:30627 |
IUPAC Name | trioxomolybdenum |
SMILES | O=[Mo](=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
13266319
|
Thermo Scientific Acros
447401000 |
100 g | Plastic bottle |
50.75 €
100g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
13276319
|
Thermo Scientific Acros
447405000 |
500 g | Plastic bottle |
167.84 €
500g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
1313-27-5 | |
143.95 | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
14802 | |
trioxomolybdenum |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
CHEBI:30627 | |
O=[Mo](=O)=O |
Specifications
1155.0°C | |
1313-27-5 | |
MoO3 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
trioxomolybdenum | |
14802 | |
143.94 | |
ACS Reagent |
795.0°C | |
MoO3 | |
MFCD00003469 | |
Solubility in water: 0.5g/L (20°C). Other solubilities: soluble in conc. mineral acids, sol. of alkali, hydroxides and ammonia, soluble in potassium bitartrate | |
O=[Mo](=O)=O | |
143.95 | |
CHEBI:30627 | |
≥99.5% | |
Molybdenum(VI) oxide |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
- Molybdenum(VI) oxide
Signal Word
- Warning
Hazard Category
- Carcinogenicity Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H351-Suspected of causing cancer.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
RUO – Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title