missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdenum(VI) Oxide, 99.5%, Honeywell Fluka™
Puriss. p.a., 99.5%
Chemical Identifiers
CAS | 1313-27-5 |
---|---|
Molecular Formula | MoO3 |
Molecular Weight (g/mol) | 143.95 |
MDL Number | MFCD00003469 |
InChI Key | JKQOBWVOAYFWKG-UHFFFAOYSA-N |
Synonym | molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide |
PubChem CID | 14802 |
ChEBI | CHEBI:30627 |
IUPAC Name | trioxomolybdenum |
SMILES | O=[Mo](=O)=O |
Description
- Leading the Industry For Over 65 Years
- Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
Chemical Identifiers
1313-27-5 | |
143.95 | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
14802 | |
trioxomolybdenum |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
CHEBI:30627 | |
O=[Mo](=O)=O |
Specifications
795°C | |
White to Yellow | |
MoO3 | |
MFCD00003469 | |
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide | |
O=[Mo](=O)=O | |
143.95 | |
CHEBI:30627 | |
99.5% | |
4.70g/cm3 |
1313-27-5 | |
Powder | |
MoO3 | |
NONH for all modes of transport | |
JKQOBWVOAYFWKG-UHFFFAOYSA-N | |
trioxomolybdenum | |
14802 | |
143.94g/mol | |
Odorless | |
Molybdenum(VI) oxide |
Safety and Handling
P261-P281-P305 + P351 + P338
missing translation for 'rtecsNumber' : 215-204-7