missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

71.96 € - 686.42 €

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11311898
SDS View Documents
Thermo Scientific Alfa Aesar
011837.22
100 g
71.96 €
100g
Please to purchase this item. Need a web account? Register with us today!
11321898
SDS View Documents
Thermo Scientific Alfa Aesar
011837.36
500 g
235.17 €
500g
Please to purchase this item. Need a web account? Register with us today!
11331898
SDS View Documents
Thermo Scientific Alfa Aesar
011837.A3
2 kg
686.42 €
2kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Molybdenum(VI) oxide is a very promising and stable molybdenum oxide for the electrochromic applications and used as a catalyst in oil industries. It is also used to make metal molybdenum. It has a wide band gap n-type semiconductor with distinctive electrochromic, thermochromic, and photochromic properties. It has been widely investigated as a smart material for catalysis sensors, lubricants, lithium battery, organic solar cells, and display materials. Molybdenum(VI) oxide (MoO3) is employed as an adhesive between enamels and metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling
Show More
SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Molybdenum(VI) oxide, 99.95% (metals basis), Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.