missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum(VI) oxide, Puratronic™, 99.998% (metals basis excluding W), W 300ppm max, Thermo Scientific Chemicals

196.98 € - 567.83 €

Chemical Identifiers

CAS 1313-27-5
Molecular Formula MoO3
Molecular Weight (g/mol) 143.95
MDL Number MFCD00003469
InChI Key JKQOBWVOAYFWKG-UHFFFAOYSA-N
Synonym molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
PubChem CID 14802
ChEBI CHEBI:30627
IUPAC Name trioxomolybdenum
SMILES O=[Mo](=O)=O
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376927
View Documents
Thermo Scientific Alfa Aesar
010812.14
25 g
196.98 €
25g
Please to purchase this item. Need a web account? Register with us today!
11386927
View Documents
Thermo Scientific Alfa Aesar
010812.22
100 g
567.83 €
100g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Its chief application is as an oxidation catalyst and as a raw material for the production of molybdenum metal. It is a Precursor to LAMOX fast ion conductors and superconductors. Molybdenum trioxide has also been suggested as a potential anti-microbial agent, e.g., in polymers. In contact with water, it forms H+ ions that can kill bacteria effectively.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

1313-27-5
143.95
JKQOBWVOAYFWKG-UHFFFAOYSA-N
14802
trioxomolybdenum
MoO3
MFCD00003469
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
CHEBI:30627
O=[Mo](=O)=O

Specifications

1155°C (sublimation)
1313-27-5
Powder
MFCD00003469
14,6239
Soluble at 0.490g/1000 cc water @ 28°C. Soluble in concentrated mineral acids.
O=[Mo](=O)=O
143.95
CHEBI:30627
99.998%
Odorless
(metals basis excluding W), W 300ppm max.
795°C
White to Gray
MoO3
UN3288
molybdenum trioxide, molybdenum vi oxide, molybdic anhydride, molybdic trioxide, molybdena, molybdic oxide, molybdenum oxide, molybdic acid anhydride, molybdenum peroxide
JKQOBWVOAYFWKG-UHFFFAOYSA-N
trioxomolybdenum
14802
143.94
Puratronic™
4.692 g/mL
Molybdenum(VI) oxide
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H300-H351-H319-H335
Fatal if swallowed.
Suspected of causing cancer.
Causes serious eye irritation.
May cause respiratory irritation.

P201-P202-P261-P264b-P271-P280i-P281-P304+P340-P305+P351+P338-P308+P313-P501c

H319-H335-H351

missing translation for 'dotInformation' : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLID, INORGANIC, N.O.S.

missing translation for 'einecsNumber' : 215-204-7

missing translation for 'rtecsNumber' : QA4725000

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.