missing translation for 'onlineSavingsMsg'
Learn More

Molybdenum wire, platinum coated, 0.457mm (0.018in) dia, Thermo Scientific Chemicals

Chemical Identifiers

CAS 7439-98-7
Molecular Formula Mo
Molecular Weight (g/mol) 95.95
MDL Number MFCD00003465
InChI Key ZOKXTWBITQBERF-UHFFFAOYSA-N
Synonym molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
PubChem CID 23932
ChEBI CHEBI:28685
IUPAC Name molybdenum
SMILES [Mo]
View More Specs

Products 3
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11376968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014658.BU
25 cm N/A
N/A
11356968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014658.G1
1 m N/A
N/A
11366968
View Documents Promotion Details
Thermo Scientific Alfa Aesar
014658.G5
5 m N/A
N/A
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

7439-98-7
95.95
ZOKXTWBITQBERF-UHFFFAOYSA-N
23932
molybdenum
Mo
MFCD00003465
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
CHEBI:28685
[Mo]

Specifications

7439-98-7
MFCD00003465
ZOKXTWBITQBERF-UHFFFAOYSA-N
molybdenum
23932
thickness 2 Micron
Mo
molybdenum, elemental, 6+ ion, mchvl, vi ion, molybdenum, metallic, metco 63, atom, molybdenum, ion 6+, tsm1, amperit 106.2
[Mo]
95.95
CHEBI:28685
Molybdenum wire, platinum coated, 0.457mm (0.018 in.) dia.
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 231-107-2

missing translation for 'tsca' : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only