missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Molybdic acid, ACS, MoO{3} 85% min, Thermo Scientific Chemicals
Click to view available options
Quantity:
500 g
2 kg
Description
Molybdic acid is used as a precursor in the production of ammonium molybdate tetrahydrate and molybdenum(IV) sulfide. The mixture of molybdic acid in concentrated sulfuric acid is called as Froehde reagent, which finds application in analytical chemistry for the detection of alkaloids.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Physical Form | Crystalline Powder |
Quantity | 2 kg |
Solubility Information | Soluble in dilute ammonium hydroxide,alkali and ammonia. Slightly soluble in water. |
Grade | ACS |
Concentration or Composition (by Analyte or Components) | MoO3 85% min. |
Odor | Odorless |
Density | 3.1g/mL |
Chemical Name or Material | Molybdic acid |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'