Learn More
Molybdic acid, ACS reagent, Thermo Scientific Chemicals
Chemical Identifiers
CAS | 7782-91-4 |
---|---|
Molecular Formula | H2MoO4 |
Molecular Weight (g/mol) | 161.96 |
MDL Number | MFCD00036287 |
InChI Key | VLAPMBHFAWRUQP-UHFFFAOYSA-L |
Synonym | molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 |
PubChem CID | 82208 |
ChEBI | CHEBI:25371 |
IUPAC Name | dihydroxy(dioxo)molybdenum |
SMILES | O[Mo](O)(=O)=O |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10200041
|
Thermo Scientific Acros
423981000 |
100 g | Plastic bottle |
N/A
|
N/A
|
||||
10197700
|
Thermo Scientific Acros
423985000 |
500 g | Plastic bottle |
N/A
|
N/A | ||||
13418578
|
Thermo Scientific Acros
423980025 |
2.5 kg | Plastic bottle |
N/A
|
N/A | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagentChemical Identifiers
7782-91-4 | |
161.96 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
82208 | |
dihydroxy(dioxo)molybdenum |
H2MoO4 | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
CHEBI:25371 | |
O[Mo](O)(=O)=O |
Specifications
7782-91-4 | |
Crystalline Powder and/or Crystals | |
MFCD00036287 | |
molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2 | |
VLAPMBHFAWRUQP-UHFFFAOYSA-L | |
dihydroxy(dioxo)molybdenum | |
82208 | |
161.96 | |
ACS Reagent | |
3.1000g/mL |
White | |
H2MoO4 | |
15,6326 | |
Solubility in water: insoluble. Other solubilities: soluble in nh4oh, h2so4 and fixed alkalies | |
O[Mo](O)(=O)=O | |
161.96 | |
CHEBI:25371 | |
≥85.0% (MoO3) | |
3.1 | |
Molybdic acid, For Analysis ACS |
Safety and Handling
GHS H Statement
Causes serious eye irritation.
May cause damage to organs through prolonged or repeated exposure.
May cause respiratory irritation.
GHS P Statement
Wear eye protection/face protection.
If eye irritation persists: Get medical advice/attention.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Call a POISON CENTER or doctor/
GHS Signal Word: Warning
missing translation for 'einecsNumber' : 231-970-5
- ''Molybdic acid''
Signal Word
- Warning
Hazard Category
- Skin corrosion/irritation Category 2
- Serious eye damage/eye irritation Category 2
- Specific target organ toxicity Category 3
Hazard Statement
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H335-May cause respiratory irritation.
Precautionary Statement
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P312-Call a POISON CENTER/doctor/if you feel unwell.
RUO – Research Use Only