missing translation for 'onlineSavingsMsg'
Learn More

Molybdic acid, for analysis, Thermo Scientific Chemicals

Product Code. 10686982
Click to view available options
Quantity:
100 g
500 g
Packaging:
Plastic bottle
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Oxidation reagent
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7782-91-4
Molecular Formula H2MoO4
Molecular Weight (g/mol) 161.96
MDL Number MFCD00036287
InChI Key VLAPMBHFAWRUQP-UHFFFAOYSA-L
Synonym molybdic acid, molybdic vi acid, unii-i96n991j1n, dihydroxy dioxo molybdenum, molybdic acid acs, dihydroxy-dioxomolybdenum, molybdate moo42-, dihydrogen, t-4, dihydroxidodioxidomolybdenum, aronis24112, moo2 oh 2
PubChem CID 82208
ChEBI CHEBI:25371
IUPAC Name dihydroxy(dioxo)molybdenum
SMILES O[Mo](O)(=O)=O

Specifications

Color White through Yellow
Physical Form Powder
Quantity 500 g
Merck Index 15, 6326
Solubility Information Solubility in water: insoluble. Other solubilities: soluble in nh4oh,h2so4 and fixed alkalies
Formula Weight 161.96
Percent Purity ≥85% (MoO3)
Grade Analytical
Packaging Plastic bottle
Specific Gravity 3.1
Density 3.1000g/mL
Chemical Name or Material Molybdic acid, p.a.
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'