Learn More
Molymod™ Molecular Model Advanced Level Chemistry Set
Brand: Molymod MMS-002
This item is not returnable.
View return policy
Specifications
Advanced Level Chemistry Set | |
18 hydrogen (l), 8 carbon (lV), 7 oxygen (ll, lV), 4 nitrogen (lll, lV), 6 chlorine (l), 6 fluorine (l), 3 sulphur (ll, lV, Vl), 2 boron (lll, lV), 4 phosphorus (lll, lV, V, Vl), 6 metal (ll, lll, lV,Vl), 3 lone pair 3D lobes (beige) Links: 26 medium (grey), 5 medium (purple), 12 long flexible (grey) | |
64 Atoms |
Product Suggestions
Customers who viewed this item also viewed.
Viewing 1-4 of
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Molymod™ Molecular Model Advanced Level Chemistry Set
Spot an opportunity for improvement?Share a Content Correction