missing translation for 'onlineSavingsMsg'
Learn More
Montmorillonite K10 Product Code.: 15449019

Montmorillonite K10

Product Code. 15449019
250 g
Click to view available options
Quantity:
250 g
This item is not returnable. View return policy

Product Code. 15449019

Brand: Thermo Scientific Alfa Aesar L15160.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1318-93-0 | Al2H2O12Si4 | 360.307 g/mol

Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used for support for catalysts, preparation of clay nanopaper composites of nacre-like structure, conversion of triose sugars with alcohols to alkyl lactates and Deprotection of ketone dimethylhydrazones.

Solubility
TRUSTED_SUSTAINABILITY

Specifications

CAS 1318-93-0
Quantity 250 g
Molecular Formula Al2H2O12Si4
MDL Number MFCD00132796
Merck Index 14,6260
Synonym montmorillonite, bentolite, gelwhite l, montmorillonite ksf, montmorillonite k 10, unii-a585mn1h2l, envirobent, neokunibond, arcillite, bedelix
Solubility Information Insoluble in water.
InChI Key GUJOJGAPFQRJSV-UHFFFAOYSA-N
SMILES O.[O-2].[O-2].[O-2].O=[Si]=O.O=[Si]=O.O=[Si]=O.O=[Si]=O.[Al+3].[Al+3]
IUPAC Name dialuminum;dioxosilane;oxygen(2-);hydrate
Molecular Weight (g/mol) 360.307
PubChem CID 71586775
Odor Odorless
Chemical Name or Material Montmorillonite K10
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249506 Certificate TypeCertificate of Analysis Date29/01/2024 Catalog NumberL15160.30
Lot Number10249491 Certificate TypeCertificate of Analysis Date26/01/2024 Catalog NumberL15160.30
Lot Number10249288 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberL15160.30
Lot Number10246548 Certificate TypeCertificate of Analysis Date04/05/2023 Catalog NumberL15160.30
Lot Number10237512 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL15160.30

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Montmorillonite K10 >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.