missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MOPS (Fine White Crystals/Molecular Biology), Fisher BioReagents™
58.39 EUR valid until 2024-09-17
Use promo code "21690" to get your promotional price.
Use promo code "21690" to get your promotional price.
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass Bottle
Poly Bottle
Description
Commonly used buffering agent
This biological buffer has a usable pH range of 6.5 to 7.9.
![TRUSTED_SUSTAINABILITY](/content/dam/fishersci/glyphs/Sustain_Badge.png)
Specifications
Melting Point | 277°C |
Boiling Point | 277°C |
Color | White |
Decomposition Information | >277°C |
pH | 2.5 to 4.0 |
Physical Form | Crystals |
Quantity | 500 g |
DNase | DNase free |
Formulation | 3-(4-Morpholino) Propane Sulfonic Acid |
Assay Percent Range | ≥97 % |
Show More |
Safety and Handling
![compliance-icons](https://webshop.fishersci.com/webfiles/eu/web-flags/GHS07_Warning.png)
Product Identifier
- MOPS
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'