missing translation for 'onlineSavingsMsg'
Learn More

MOPS, MP Biomedicals™

35.75 € - 833.00 €

Chemical Identifiers

CAS 1132-61-2
Molecular Formula C7H15NO4S
Molecular Weight (g/mol) 209.26
MDL Number MFCD00006183
InChI Key DVLFYONBTKHTER-UHFFFAOYSA-N
Synonym 3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
PubChem CID 70807
ChEBI CHEBI:44115
SMILES [O-]S(=O)(=O)CCC[NH+]1CCOCC1
View More Specs

Products 4
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11473610
View Documents
MP Biomedicals
0210237010
10 g
35.75 €
10g
Please to purchase this item. Need a web account? Register with us today!
11483610
View Documents
MP Biomedicals
0210237080
100 g
143.00 €
100g
Please to purchase this item. Need a web account? Register with us today!
11493610
View Documents
MP Biomedicals
0210237083
250 g
303.00 €
250g
Please to purchase this item. Need a web account? Register with us today!
11403620
View Documents
MP Biomedicals
0210237091
1 kg
833.00 €
1kg
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Specifications

Chemical Identifiers

1132-61-2
209.26
DVLFYONBTKHTER-UHFFFAOYSA-N
70807
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
C7H15NO4S
MFCD00006183
3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
CHEBI:44115

Specifications

1132-61-2
Powder
C7H15NO4S
3-(N-morpholino)propanesulfonic acid, 4-morpholinepropanesulfonic acid
[O-]S(=O)(=O)CCC[NH+]1CCOCC1
209.26
CHEBI:44115
MOPS
White
100.4%
MFCD00006183
DVLFYONBTKHTER-UHFFFAOYSA-N
3-morpholin-4-ylpropane-1-sulfonic acid
70807
209.3
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 214-478-5

Product Identifier
  • MOPS

Signal Word
  • Warning

Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P321-Specific treatment (see on this label).
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : MOPS

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.