missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 0.2M buffer soln., pH 6.5, Thermo Scientific™

Product Code. 15474329
Click to view available options
Quantity:
100 mL
250 mL
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 79803-73-9
Molecular Formula C7H14NNaO5S
Molecular Weight (g/mol) 247.241
MDL Number MFCD00070007
InChI Key WSFQLUVWDKCYSW-UHFFFAOYSA-M
PubChem CID 23675131
IUPAC Name sodium;2-hydroxy-3-morpholin-4-ylpropane-1-sulfonate
SMILES C1COCCN1CC(CS(=O)(=O)[O-])O.[Na+]

Specifications

Concentration 0.2 M
pH 6.5
Physical Form Liquid
Quantity 100 mL
Chemical Name or Material MOPSO buffer soln.
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.