missing translation for 'onlineSavingsMsg'
Learn More

MOPSO, 98%, Thermo Scientific Chemicals

Product Code. 11309628
Click to view available options
Quantity:
10 g
50 g
250 g
1 kg
This item is not returnable. View return policy
This item is not returnable. View return policy

Biological buffers, pH range 6.2 to 7.6

It is an important raw material used intermediate used in pharmaceuticals, organic synthesis, dye stuffs and agrochemicals. Also used as biological buffers, pH range 6.2-7.6.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 68399-77-9
Molecular Formula C7H15NO5S
Molecular Weight (g/mol) 225.259
MDL Number MFCD00009629
InChI Key NUFBIAUZAMHTSP-UHFFFAOYSA-N
Synonym mopso, 2-hydroxy-3-morpholinopropanesulfonic acid, 2-hydroxy-3-morpholinopropane-1-sulfonic acid, 2-hydroxy-4-morpholinepropanesulphonic acid, 3-morpholino-2-hydroxypropanesulfonic acid, 3-n-morpholino-2-hydroxypropanesulfonic acid, 2-hydroxy-3-morpholin-4-yl propane-1-sulfonic acid, beta-hydroxy-4-morpholinepropanesulfonic acid, 3-4-morpholino-2-hydroxypropanesulfonic acid, 4-morpholinepropanesulfonic acid, .beta.-hydroxy
PubChem CID 109333
IUPAC Name 2-hydroxy-3-morpholin-4-ylpropane-1-sulfonic acid
SMILES C1COCCN1CC(CS(=O)(=O)O)O

Specifications

Melting Point 276°C to 279°C
Physical Form Powder
Quantity 250 g
Solubility Information Water solubility under desired conditions ca.112,6g/L at 20°C.
Formula Weight 225.27
Percent Purity 98%
Chemical Name or Material MOPSO

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'