missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Morpholin-2-ylmethyl-carbamic acid tert-butyl ester, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
IUPAC Name | tert-butyl N-[(morpholin-2-yl)methyl]carbamate |
Formula Weight | 216.28 |
Percent Purity | 97% |
Quantity | 5 g |
Chemical Name or Material | Morpholin-2-ylmethyl-carbamic acid tert-butyl ester |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'