missing translation for 'onlineSavingsMsg'
Learn More
Morpholine-4-carboxamidine hydrobromide, 98%, Thermo Scientific Chemicals Product Code.: 11450421

Morpholine-4-carboxamidine hydrobromide, 98%, Thermo Scientific Chemicals

Product Code. 11450421
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11450421

Brand: Thermo Scientific Alfa Aesar A19985.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

It is used as an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used as an active pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Hygroscopic. Store away from oxidizing agents, water/moisture. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 157415-17-3
Molecular Formula C5H12BrN3O
Molecular Weight (g/mol) 210.075
MDL Number MFCD00051994
InChI Key IOHSDHMCZXCESZ-UHFFFAOYSA-N
Synonym morpholine-4-carboximidamide hydrobromide, morpholinoformamidine hydrobromide, 4-morpholinylformamidine hydrobromide, morpholine-4-carboxamidine hydrobromide, 4-morpholinecarboximidamide, monohydrobromide, acmc-20adpt, morpholine-4-carboxamidine, bromide, morpholin-4-carboxamidine hydrobromide
PubChem CID 2806564
IUPAC Name morpholine-4-carboximidamide;hydrobromide
SMILES C1COCCN1C(=N)N.Br

Specifications

Melting Point ∼155°C to 165°C
Quantity 100 g
Beilstein 7389038
Sensitivity Hygroscopic
Solubility Information Soluble in water.
Formula Weight 210.08
Percent Purity 98%
Chemical Name or Material Morpholine-4-carboxamidine hydrobromide

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
Morpholine-4-carboxamidine hydrobromide, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.